2-Methoxy-3-pyridinboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Methoxypyridine-3-boronic acid

016262-1G
FLUO016262-1GEA 34.1 EUR
FLUO016262-1G
2-Methoxy-3-pyridinboronsäure
2-Methoxy-3-pyridineboronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152,95 g/mol
MDL: MFCD03411572
CAS-Nummer: 163105-90-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR