2-Nitrophenylboronsäure

Lieferant: FLUOROCHEM

016265-1G 016265-5G
FLUO016265-1GEA 48.8 EUR
FLUO016265-1G FLUO016265-5G
2-Nitrophenylboronsäure
2-Nitrophenylboronsäure
Formel: C₆H₆BNO₄
MDL: MFCD00161358
CAS-Nummer: 5570-19-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR