2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid

Lieferant: FLUOROCHEM

CFF96.1 017084-2.5G
CHMPCFF96.1EA 508 EUR
CHMPCFF96.1 FLUO017084-2.5G
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
2-Methyl-6-(trifluormethyl)nicotinsäurehydrazid
Formel: C₈H₈F₃N₃O
MDL: MFCD01565843
CAS-Nummer: 402479-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR