4-Amino-6-bromchinolin

Lieferant: FLUOROCHEM

CFF749.1 017128-1G
CHMPCFF749.1EA 522 EUR
CHMPCFF749.1 FLUO017128-1G
4-Amino-6-bromchinolin
4-Amino-6-bromoquinoline
Formel: C₉H₇BrN₂
Molecular Weight: 223,0693 g/mol
MDL: MFCD03407953
CAS-Nummer: 65340-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR