2-Chlor-3-fluorbenzamid

Lieferant: FLUOROCHEM

017140-1G
FLUO017140-1GEA 174 EUR
FLUO017140-1G
2-Chlor-3-fluorbenzamid
2-Chlor-3-fluorbenzamid
Formel: C₇H₅ClFNO
CAS-Nummer: 683274-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR