1-(2-Chlorphenyl)biguanid Hydrochlorid

Lieferant: FLUOROCHEM

017144-1G
FLUO017144-1GEA 0 EUR
FLUO017144-1G
1-(2-Chlorphenyl)biguanid Hydrochlorid
1-(2-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248.12 g/mol
MDL: MFCD00053016
CAS-Nummer: 19579-44-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR