1-(3-Chlorphenyl)biguanid Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: m-CPBG hydrochloride

017145-1G
FLUO017145-1GEA 0 EUR
FLUO017145-1G
1-(3-Chlorphenyl)biguanid Hydrochlorid
1-(3-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248,12 g/mol
MDL: MFCD00053019
CAS-Nummer: 2113-05-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR