1-(4-Chlorphenyl)biguanid Hydrochlorid

Lieferant: FLUOROCHEM

017146-1G
FLUO017146-1GEA 0 EUR
FLUO017146-1G
1-(4-Chlorphenyl)biguanid Hydrochlorid
1-(4-Chlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₁Cl₂N₅
Molecular Weight: 248,12 g/mol
MDL: MFCD00053020
CAS-Nummer: 4022-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR