3-Chlor-6-hydrazinopyridazin
Lieferant: FLUOROCHEM
Synonyme:
3-Chloro-6-hydrazinylpyridazine
CFF984.
017148-1G
CHMPCFF984.EA
0
EUR
CHMPCFF984.
FLUO017148-1G
3-Chlor-6-hydrazinopyridazin
3-Chlor-6-hydrazinopyridazin
Formel:
C₄H₅ClN₄ Molecular Weight: 144,56 g/mol |
MDL:
MFCD00051740 CAS-Nummer: 17284-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...