4-(4-Fluor-2-nitrophenyl)morpholin

Lieferant: FLUOROCHEM

017159-1G 017159-5G
FLUO017159-1GEA 76.4 EUR
FLUO017159-1G FLUO017159-5G
4-(4-Fluor-2-nitrophenyl)morpholin
4-(4-Fluor-2-nitrophenyl)morpholin
Formel: C₁₀H₁₁FN₂O₃
Molecular Weight: 226.20 g/mol
MDL: MFCD00798369
CAS-Nummer: 238418-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR