1-(4-Fluor-2-nitrophenyl)piperidin
Lieferant: FLUOROCHEM
CFF1140.1
017160-1G
CHMPCFF1140.1EA
522
EUR
CHMPCFF1140.1
FLUO017160-1G
1-(4-Fluor-2-nitrophenyl)piperidin
1-(4-Fluor-2-nitrophenyl)piperidin
Formel:
C₁₁H₁₃FN₂O₂ Molecular Weight: 224,23 g/mol |
MDL:
MFCD00798370 CAS-Nummer: 719-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...