1-(4-Fluor-2-nitrophenyl)pyrrolidin

Lieferant: FLUOROCHEM

CFF1141.1 017161-1G
CHMPCFF1141.1EA 522 EUR
CHMPCFF1141.1 FLUO017161-1G
1-(4-Fluor-2-nitrophenyl)pyrrolidin
1-(4-Fluor-2-nitrophenyl)pyrrolidin
Formel: C₁₀H₁₁ClN₂O₂
Molecular Weight: 210,20 g/mol
MDL: MFCD00798372
CAS-Nummer: 778-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR