3-Brom-2-thiophencarbaldehyd
Lieferant: FLUOROCHEM
Synonyme:
3-Bromothiophene-2-carboxaldehyde, 3-Bromo-2-formylthiophene
017210-2.5G
FLUO017210-2.5GEA
0
EUR
FLUO017210-2.5G
3-Brom-2-thiophencarbaldehyd
3-Brom-2-thiophencarbaldehyd
Formel:
C₅H₃BrOS Molecular Weight: 191.05 g/mol |
MDL:
MFCD00126680 CAS-Nummer: 930-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...