3-Aminoisonicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid
017211-1G
FLUO017211-1GEA
31.1
EUR
FLUO017211-1G
3-Aminoisonicotinsäure
3-Aminoisonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol |
MDL:
MFCD00137840 CAS-Nummer: 7579-20-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...