3,4-Diaminobenzonitril

Lieferant: FLUOROCHEM

017243-1G
FLUO017243-1GEA 34.1 EUR
FLUO017243-1G
3,4-Diaminobenzonitril
3,4-Diaminobenzonitril
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
MDL: MFCD00723901
CAS-Nummer: 17626-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR