3-(Dimethylamino)benzamid

Lieferant: FLUOROCHEM

CFF250.1 017301-1G
CHMPCFF250.1EA 268 EUR
CHMPCFF250.1 FLUO017301-1G
3-(Dimethylamino)benzamid
3-(Dimethylamino)benzamid
Formel: C₉H₁₂N₂O
MDL: MFCD01196355
CAS-Nummer: 33225-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR