3-Methylbenzamidoxim

Lieferant: FLUOROCHEM

Synonyme: N-Hydroxy-3-methylbenzimidamide

CFF268.5 CFF268.25 017315-5G 017315-10G
CHMPCFF268.5EA 532 EUR
CHMPCFF268.5 CHMPCFF268.25 FLUO017315-5G FLUO017315-10G
3-Methylbenzamidoxim
3-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
Molecular Weight: 150.18 g/mol
MDL: MFCD00833266
CAS-Nummer: 40067-82-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR