N-Boc-ethylendiamin

Lieferant: FLUOROCHEM

Synonyme: t-boc-N-EDA

CFF275.2.5 CFF275.10 017322-2.5G 017322-10G 017322-25G 017322-5G
CHMPCFF275.2.5EA 348 EUR
CHMPCFF275.2.5 CHMPCFF275.10 FLUO017322-2.5G FLUO017322-10G FLUO017322-25G FLUO017322-5G
N-Boc-ethylendiamin
N-Boc-ethylendiamin
Formel: C₇H₁₆N₂O₂
MDL: MFCD00191871
CAS-Nummer: 57260-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR