2,6-Dimethoxyanilin
Lieferant: FLUOROCHEM
CFF294.1
017336-1G
CHMPCFF294.1EA
348
EUR
CHMPCFF294.1
FLUO017336-1G
2,6-Dimethoxyanilin
2,6-Dimethoxyanilin
Formel:
C₈H₁₁NO₂ |
MDL:
MFCD00053934 CAS-Nummer: 2734-70-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...