3-(Chlormethyl)-1,2,4-oxadiazol

Lieferant: FLUOROCHEM

CFF295.5 017337-5G 017337-1G
CHMPCFF295.5EA 613 EUR
CHMPCFF295.5 FLUO017337-5G FLUO017337-1G
3-(Chlormethyl)-1,2,4-oxadiazol
3-(Chloromethyl)-1,2,4-oxadiazole
Formel: C₃H₃ClN₂O
MDL: MFCD00052097
CAS-Nummer: 51791-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR