1-(2-Cyanphenyl)piperidin
Lieferant: FLUOROCHEM
Synonyme:
2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile
CFF300.5
017342-5G
CHMPCFF300.5EA
268
EUR
CHMPCFF300.5
FLUO017342-5G
1-(2-Cyanphenyl)piperidin
1-(2-Cyanphenyl)piperidin
Formel:
C₁₂H₁₄N₂ Molecular Weight: 186.26 g/mol |
MDL:
MFCD00049221 CAS-Nummer: 72752-52-4 EINECS: 427-330-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...