5-Isoxazolcarbonylchlorid

Lieferant: FLUOROCHEM

CFF304.5 CFF304.25 017346-5G
CHMPCFF304.5EA 508 EUR
CHMPCFF304.5 CHMPCFF304.25 FLUO017346-5G
5-Isoxazolcarbonylchlorid
5-Isoxazolcarbonylchlorid
Formel: C₄H₂ClNO₂
Molecular Weight: 131,52 g/mol
Siedepunkt: 74…76 °C (1013 hPa)
MDL: MFCD00067868
CAS-Nummer: 62348-13-4
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR