2-Cyanbenzolsulfonylchlorid

Lieferant: FLUOROCHEM

Synonyme: 2-Cyanbenzol-1-sulfonylchlorid, 2-Cyanbenzolsulfonsäurechlorid, 2-Cyanbenzolsulfochlorid

CFF309.2.5 CFF309.10 CFF309.25 017349-25G 017349-1G 017349-5G
CHMPCFF309.2.5EA 268 EUR
CHMPCFF309.2.5 CHMPCFF309.10 CHMPCFF309.25 FLUO017349-25G FLUO017349-1G FLUO017349-5G
2-Cyanbenzolsulfonylchlorid
2-Cyanbenzolsulfonylchlorid
Formel: C₇H₄ClNO₂S
Molecular Weight: 201.63 g/mol
Schmelzpunkt: 67,5
MDL: MFCD00052341
CAS-Nummer: 69360-26-5
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR