2,4-Dimethyl-5-thiazolcarbonsäure

Lieferant: FLUOROCHEM

CFF387.5 CFF387.10 CFF387.25 017412-5G 017412-10G 017412-25G 017412-1G
CHMPCFF387.5EA 375 EUR
CHMPCFF387.5 CHMPCFF387.10 CHMPCFF387.25 FLUO017412-5G FLUO017412-10G FLUO017412-25G FLUO017412-1G
2,4-Dimethyl-5-thiazolcarbonsäure
2,4-Dimethyl-5-thiazolecarboxylic acid
Formel: C₆H₇NO₂S
MDL: MFCD00052943
CAS-Nummer: 53137-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR