1,4-Benzodioxan-2-carbonylchlorid

Lieferant: FLUOROCHEM

017521-1G 017521-5G
FLUO017521-1GEA 134 EUR
FLUO017521-1G FLUO017521-5G
1,4-Benzodioxan-2-carbonylchlorid
1,4-Benzodioxan-2-carbonylchlorid
Formel: C₉H₇ClO₃
MDL: MFCD02180342
CAS-Nummer: 3663-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR