4-(4-Methylphenyl)-3-thiosemicarbazid

Lieferant: FLUOROCHEM

Synonyme: N-(p-Tolyl)hydrazinecarbothioamide

018014-5G
FLUO018014-5GEA 130 EUR
FLUO018014-5G
4-(4-Methylphenyl)-3-thiosemicarbazid
4-(4-Methylphenyl)-3-thiosemicarbazid
Formel: C₈H₁₁N₃S
Molecular Weight: 181,26 g/mol
MDL: MFCD00041302
CAS-Nummer: 13278-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR