4-Fluor-3-nitrobenzoesäure
Lieferant: FLUOROCHEM
|
Achtung
|
018022-100G
FLUO018022-100GEA
135
EUR
FLUO018022-100G
4-Fluor-3-nitrobenzoesäure
4-Fluoro-3-nitrobenzoic acid
Formel:
C₇H₄FNO₄ Molecular Weight: 185.11 g/mol Siedepunkt: 92 °C (15 mmHg) Schmelzpunkt: 123…124 °C |
MDL:
MFCD00007058 CAS-Nummer: 453-71-4 EINECS: 207-221-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...