3-(4-Bromphenyl)propiononitril
Lieferant: FLUOROCHEM
Synonyme:
3-(4-Bromophenyl)propanenitrile
018025-1G
FLUO018025-1GEA
91.5
EUR
FLUO018025-1G
3-(4-Bromphenyl)propiononitril
3-(4-Bromphenyl)propiononitril
Formel:
C₉H₈BrN Molecular Weight: 210,0705 g/mol |
MDL:
MFCD01310795 CAS-Nummer: 57775-08-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...