1-(3-Methyl-2-pyridinyl)piperazin
Lieferant: FLUOROCHEM
018129-1G
FLUO018129-1GEA
359
EUR
FLUO018129-1G
1-(3-Methyl-2-pyridinyl)piperazin
1-(3-Methyl-2-pyridinyl)piperazin
Formel:
C₁₀H₁₅N₃ |
MDL:
MFCD03412117 CAS-Nummer: 104396-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...