4-Acetoxybenzoesäure

Lieferant: FLUOROCHEM

018147-10G
FLUO018147-10GEA 0 EUR
FLUO018147-10G
4-Acetoxybenzoesäure
4-Acetoxybenzoesäure
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00002540
CAS-Nummer: 2345-34-8
EINECS: 219-066-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR