1-(3-Acetylphenyl)-2-thioharnstoff

Lieferant: FLUOROCHEM

018155-10G
FLUO018155-10GEA 372 EUR
FLUO018155-10G
1-(3-Acetylphenyl)-2-thioharnstoff
1-(3-Acetylphenyl)-2-thioharnstoff
Formel: C₉H₁₀N₂OS
MDL: MFCD00041170
CAS-Nummer: 86801-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR