4-Allyl-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018164-10G
FLUO018164-10GEA 162 EUR
FLUO018164-10G
4-Allyl-3-thiosemicarbazid
4-Allyl-3-thiosemicarbazid
Formel: C₄H₉N₃S
MDL: MFCD00007619
CAS-Nummer: 3766-55-0
EINECS: 223-186-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR