3-Aminobenzamid

Lieferant: FLUOROCHEM

Synonyme: 3-Aminobenzoesäureamid

018167-5G
FLUO018167-5GEA 84.9 EUR
FLUO018167-5G
3-Aminobenzamid
3-Aminobenzamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Siedepunkt: 341 °C (1013 hPa)
Schmelzpunkt: 111…115 °C
MDL: MFCD00007989
CAS-Nummer: 3544-24-9
EINECS: 222-586-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR