4-Aminobenzamid

Lieferant: FLUOROCHEM

Synonyme: 4-Aminobenzoesäureamid

018168-25G
FLUO018168-25GEA 32.1 EUR
FLUO018168-25G
4-Aminobenzamid
4-Aminobenzamid
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007999
CAS-Nummer: 2835-68-9
EINECS: 220-612-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR