3-Amino-5-tert-butylisoxazol

Lieferant: FLUOROCHEM

018173-5G
FLUO018173-5GEA 51.6 EUR
FLUO018173-5G
3-Amino-5-tert-butylisoxazol
3-Amino-5-tert-butylisoxazol
Formel: C₇H₁₂N₂O
Molecular Weight: 140,19 g/mol
MDL: MFCD00055620
CAS-Nummer: 55809-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR