5-Amino-3-methylisoxazol
Lieferant: FLUOROCHEM
Synonyme:
3-Methylisoxazol-5-ylamine, 3-Methylisoxazol-5-amine
018187-5G
FLUO018187-5GEA
99.3
EUR
FLUO018187-5G
5-Amino-3-methylisoxazol
5-Amino-3-methylisoxazole
Formel:
C₄H₆N₂O Molecular Weight: 98,1 g/mol Siedepunkt: 194 °C (1013 hPa) Schmelzpunkt: 84 °C |
MDL:
MFCD00003151 CAS-Nummer: 14678-02-5 EINECS: 238-719-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...