Benzhydrylisothiocyanat

Lieferant: FLUOROCHEM

018194-5G
FLUO018194-5GEA 72.3 EUR
FLUO018194-5G
Benzhydrylisothiocyanat
Benzhydrylisothiocyanat
Formel: C₁₄H₁₁NS
Molecular Weight: 225,31 g/mol
MDL: MFCD00046815
CAS-Nummer: 3550-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR