4-Benzhydryl-3-thiosemicarbazid

Lieferant: FLUOROCHEM

018195-1G
FLUO018195-1GEA 134 EUR
FLUO018195-1G
4-Benzhydryl-3-thiosemicarbazid
4-Benzhydryl-3-thiosemicarbazid
Formel: C₁₄H₁₅N₃S
MDL: MFCD00060586
CAS-Nummer: 21198-25-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR