Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)

Lieferant: FLUOROCHEM

Synonyme: Benzo-18-crown 6-Ether

018197-1G
FLUO018197-1GEA 115 EUR
FLUO018197-1G
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Formel: C₁₆H₂₄O₆
Molecular Weight: 312.36 g/mol
MDL: MFCD00062741
CAS-Nummer: 14098-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR