4-Benzylanilin
Lieferant: FLUOROCHEM
Synonyme:
4-Aminodiphenylmethane
018207-5G
FLUO018207-5GEA
84.9
EUR
FLUO018207-5G
4-Benzylanilin
4-Benzylanilin
Formel:
C₁₃H₁₃N Molecular Weight: 183,25 g/mol Siedepunkt: 300 °C (1013 hPa) Schmelzpunkt: 35…36 °C Dichte: 1,07 g/cm³ (20 °C) |
MDL:
MFCD00047861 CAS-Nummer: 1135-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...