Benzylcarbazat

Lieferant: FLUOROCHEM

Synonyme: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate

018210-5G
FLUO018210-5GEA 0 EUR
FLUO018210-5G
Benzylcarbazat
Benzylcarbazat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
MDL: MFCD00041890
CAS-Nummer: 5331-43-1
EINECS: 226-230-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR