Benzyl-4-hydroxybenzoat

Lieferant: FLUOROCHEM
Achtung

Synonyme: Benzylparaben, 4-Hydroxybenzoesäurebenzylester

018211-10G
FLUO018211-10GEA 0 EUR
FLUO018211-10G
Benzyl-4-hydroxybenzoat
Benzyl-4-hydroxybenzoat
Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
MDL: MFCD00016471
CAS-Nummer: 94-18-8
EINECS: 202-311-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR