4-Benzyloxyanisol
Lieferant: FLUOROCHEM
018215-10G
FLUO018215-10GEA
0
EUR
FLUO018215-10G
4-Benzyloxyanisol
4-Benzyloxyanisol
Formel:
C₁₄H₁₄O₂ |
MDL:
MFCD00025806 CAS-Nummer: 6630-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...