1-Benzyloxy-4-iodbenzol

Lieferant: FLUOROCHEM

Synonyme: 1-(Benzyloxy)-4-iodobenzene, 4-Benzyloxyiodobenzene

018223-5G
FLUO018223-5GEA 55.4 EUR
FLUO018223-5G
1-Benzyloxy-4-iodbenzol
1-Benzyloxy-4-iodbenzol
Formel: C₁₃H₁₁IO
Molecular Weight: 310,13 g/mol
MDL: MFCD00079704
CAS-Nummer: 19578-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR