(2-Benzyloxyphenyl)essigsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(2-(Benzyloxy)phenyl)acetic acid

018225-1G
FLUO018225-1GEA 152 EUR
FLUO018225-1G
(2-Benzyloxyphenyl)essigsäure
(2-Benzyloxyphenyl)essigsäure
Formel: C₆H₅CH₂OC₆H₄CH₂COOH
Molecular Weight: 242.27 g/mol
MDL: MFCD00016822
CAS-Nummer: 22047-88-7
UN: 3077
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR