4-Benzyl-3-thiosemicarbazid
Lieferant: FLUOROCHEM
Synonyme:
N-Benzylhydrazinecarbothioamide
018233-10G
FLUO018233-10GEA
372
EUR
FLUO018233-10G
4-Benzyl-3-thiosemicarbazid
4-Benzyl-3-thiosemicarbazid
Formel:
C₈H₁₁N₃S Molecular Weight: 181,26 g/mol |
MDL:
MFCD00025145 CAS-Nummer: 13431-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...