2-Biphenylisothiocyanat
Lieferant: FLUOROCHEM
018239-5G
FLUO018239-5GEA
360
EUR
FLUO018239-5G
2-Biphenylisothiocyanat
2-Biphenylisothiocyanat
Formel:
C₁₃H₉NS Molecular Weight: 211,29 g/mol |
MDL:
MFCD00022052 CAS-Nummer: 19394-61-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...