3-Brombenzhydrazid
Lieferant: FLUOROCHEM
Synonyme:
3-Bromobenzoic hydrazide
018243-10G
FLUO018243-10GEA
85.1
EUR
FLUO018243-10G
3-Brombenzhydrazid
3-Brombenzhydrazid
Formel:
C₇H₇BrN₂O Molecular Weight: 215,05 g/mol Siedepunkt: 366 °C (1013 hPa) Schmelzpunkt: 151 °C |
MDL:
MFCD00007600 CAS-Nummer: 39115-96-3 EINECS: 254-298-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...