3-Brom-4-chlorbenzhydrazid

Lieferant: FLUOROCHEM

018251-1G
FLUO018251-1GEA 257 EUR
FLUO018251-1G
3-Brom-4-chlorbenzhydrazid
3-Brom-4-chlorbenzhydrazid
Formel: C₇H₆BrClN₂O
MDL: MFCD00270094
CAS-Nummer: 148993-18-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR