4-Brom-2-chlorbenzhydrazid

Lieferant: FLUOROCHEM

018252-1G
FLUO018252-1GEA 306 EUR
FLUO018252-1G
4-Brom-2-chlorbenzhydrazid
4-Brom-2-chlorbenzhydrazid
Formel: C₇H₆BrClN₂O
MDL: MFCD00270095
CAS-Nummer: 206559-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR